Assay Details
Target Gene Details
Entrez Gene ID: | 2717 |
Gene Name: | galactosidase alpha |
Gene Aliases: |
GALA |
Location: |
Chr.X:101397791-101408013 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLA | NM_000169.2 | NP_000160.1 | ||
| AK222627.1 | BAD96347.1 | |||
| AK291095.1 | ||||
| BC002689.2 | AAH02689.1 | |||
| BT006864.1 | AAP35510.1 | |||
| D00039.1 | BAA34059.1 | |||
| DB370480.1 | ||||
| X05790.1 | CAA29232.1 | |||
| X16889.1 |
Target Gene Details
Entrez Gene ID: | 100529097 |
Gene Name: | RPL36A-HNRNPH2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:101390890-101414140 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL36A-HNRNPH2 | NM_001199973.1 | NP_001186902.1 | ||
| NM_001199974.1 | NP_001186903.1 | |||
| DA530036.1 | ||||
| DA998453.1 |
More Information
Set Membership: |
|

Genomic Map