Assay Details
Target Gene Details
Entrez Gene ID: | 401612 |
Gene Name: | solute carrier family 25 member 53 |
Gene Aliases: |
MCART6 |
Location: |
Chr.X:104099214-104157027 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A53 | NM_001012755.4 | NP_001012773.2 | ||
| XM_005262129.4 | XP_005262186.1 | |||
| XM_011530952.2 | XP_011529254.1 | |||
| XM_011530953.2 | XP_011529255.1 | |||
| XM_017029514.1 | XP_016885003.1 | |||
| XM_017029515.1 | XP_016885004.1 | |||
| XM_017029516.1 | XP_016885005.1 | |||
| AL833609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469772 | Chr.X:103928672 - 104131003 on Build GRCh38 | Gain |
|
| esv2758880 | Chr.X:103827471 - 104210380 on Build GRCh38 | Gain |
|
| nsv519626 | Chr.X:103933493 - 104158390 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map