Assay Details
Target Gene Details
Entrez Gene ID: | 101928650 |
Gene Name: | uncharacterized LOC101928650 |
Gene Aliases: |
- |
Location: |
Chr.1:170461317-170532609 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928650 | NR_125958.1 | |||
| AK096329.1 | ||||
| HY028703.1 | ||||
| HY035829.1 |

Genomic Map