Assay Details
Target Gene Details
Entrez Gene ID: | 101929147 |
Gene Name: | uncharacterized LOC101929147 |
Gene Aliases: |
- |
Location: |
Chr.1:119140396-119275973 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929147 | NR_125974.1 | |||
| NR_125975.1 | ||||
| NR_125976.1 | ||||
| AK125759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831192 | Chr.1:119128549 - 119300603 on Build GRCh38 | Gain |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74592153] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map