Assay Details
Target Gene Details
Entrez Gene ID: | 59269 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 3 |
Gene Aliases: |
KBP-1, KBP1, KRC, SHN3, Schnurri-3, ZAS3, ZNF40C |
Location: |
Chr.1:41506365-42035925 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIVEP3 | NM_001127714.2 | NP_001121186.1 | ||
| NM_024503.4 | NP_078779.2 | |||
| XM_011541884.2 | XP_011540186.1 | |||
| XM_017001992.1 | XP_016857481.1 | |||
| XM_017001993.1 | XP_016857482.1 | |||
| XM_017001994.1 | XP_016857483.1 | |||
| AB046775.3 | ||||
| AF278765.1 | AAK01082.1 | |||
| AK122991.1 | ||||
| BC152563.1 | ||||
| M61743.1 | AAA36137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv633 | Chr.1:41479827 - 41524947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map