Assay Details
Target Gene Details
Entrez Gene ID: | 1806 |
Gene Name: | dihydropyrimidine dehydrogenase |
Gene Aliases: |
DHP, DHPDHASE, DPD |
Location: |
Chr.1:97077743-97921059 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYD | NM_000110.3 | NP_000101.2 | ||
| XM_005270562.3 | XP_005270619.2 | |||
| XM_017000507.1 | XP_016855996.1 | |||
| XM_017000508.1 | XP_016855997.1 | |||
| XM_017000509.1 | XP_016855998.1 | |||
| XM_017000510.1 | XP_016855999.1 | |||
| AB003063.1 | BAA89789.1 | |||
| AK291217.1 | ||||
| BC131777.1 | ||||
| BC131778.1 | ||||
| U09178.1 | AAA57474.1 | |||
| U20938.1 | AAB51366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830692 | Chr.1:97035936 - 97159597 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map