Assay Details
Target Gene Details
Entrez Gene ID: | 103752588 |
Gene Name: | PTGS2 antisense NFKB1 complex-mediated expression regulator RNA |
Gene Aliases: |
PACER, PTGS2-AS1 |
Location: |
Chr.1:186680654-186681446 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACERR | NR_125801.1 | 1 | 77 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470750 | Chr.1:186673297 - 186731827 on Build GRCh38 | Loss |
|
| esv2762854 | Chr.1:186577969 - 186878442 on Build GRCh38 | Gain |
|

Genomic Map