Assay Details
Target Gene Details
Entrez Gene ID: | 23566 |
Gene Name: | lysophosphatidic acid receptor 3 |
Gene Aliases: |
EDG7, Edg-7, GPCR, HOFNH30, LP-A3, LPA3, RP4-678I3 |
Location: |
Chr.1:84813403-84893213 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPAR3 | NM_012152.2 | NP_036284.1 | ||
| AF127138.1 | AAD56311.1 | |||
| AF186380.1 | AAF00530.1 | |||
| AF236117.1 | AAF91291.1 | |||
| AK308283.1 | ||||
| AY322547.1 | AAP84360.1 | |||
| BC126268.1 | ||||
| BC126270.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116825108] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map