Assay Details
Target Gene Details
Entrez Gene ID: | 9473 |
Gene Name: | thymocyte selection associated family member 2 |
Gene Aliases: |
C1orf38, ICB-1 |
Location: |
Chr.1:27872531-27886685 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THEMIS2 | NM_001039477.2 | NP_001034566.1 | ||
| NM_001105556.2 | NP_001099026.1 | |||
| NM_001286113.1 | NP_001273042.1 | |||
| NM_001286115.1 | NP_001273044.1 | |||
| NM_004848.3 | NP_004839.2 | |||
| XM_005246041.3 | XP_005246098.1 | |||
| XM_006711050.1 | XP_006711113.1 | |||
| XM_011542445.2 | XP_011540747.1 | |||
| AB035482.1 | BAA96464.1 | |||
| AB050854.1 | BAB33313.1 | |||
| AF044896.1 | AAC16284.1 | |||
| AK303090.1 | ||||
| AK303141.1 | ||||
| BC031655.1 | ||||
| BC132692.1 | ||||
| BC133049.1 | ||||
| BG716895.1 | ||||
| BI908174.1 | ||||
| CR749321.1 | CAH18176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509069 | Chr.1:27817460 - 27905956 on Build GRCh38 | Insertion |
|
| esv3585564 | Chr.1:27879549 - 27935475 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map