Assay Details
Target Gene Details
Entrez Gene ID: | 84871 |
Gene Name: | ATP/GTP binding protein like 4 |
Gene Aliases: |
CCP6 |
Location: |
Chr.1:48520157-50024011 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL4 | XM_017002595.1 | 13 | 3289 | XP_016858084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585970 | Chr.1:48459872 - 48603418 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map