Assay Details
Target Gene Details
Entrez Gene ID: | 387597 |
Gene Name: | immunoglobulin like domain containing receptor 2 |
Gene Aliases: |
C1orf32, dJ782G3.1 |
Location: |
Chr.1:166913204-166975324 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILDR2 | NM_199351.2 | NP_955383.1 | ||
| XM_006711314.3 | XP_006711377.1 | |||
| XM_017001252.1 | XP_016856741.1 | |||
| XM_017001253.1 | XP_016856742.1 | |||
| XM_017001254.1 | XP_016856743.1 | |||
| XM_017001255.1 | XP_016856744.1 | |||
| XM_017001256.1 | XP_016856745.1 | |||
| XM_017001257.1 | XP_016856746.1 | |||
| XM_017001258.1 | XP_016856747.1 | |||
| AF503509.1 | AAQ07593.1 |
Target Gene Details
Entrez Gene ID: | 84944 |
Gene Name: | maelstrom spermatogenic transposon silencer |
Gene Aliases: |
CT128, SPATA35 |
Location: |
Chr.1:166921121-167022214 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAEL | XM_017002602.1 | XP_016858091.1 |
More Information
Set Membership: |
|

Genomic Map