Assay Details
Target Gene Details
Entrez Gene ID: | 7520 |
Gene Name: | X-ray repair cross complementing 5 |
Gene Aliases: |
KARP-1, KARP1, KU80, KUB2, Ku86, NFIV |
Location: |
Chr.2:216109297-216206293 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XRCC5 | NM_021141.3 | NP_066964.1 | ||
| AK222603.1 | BAD96323.1 | |||
| AK290740.1 | ||||
| BC019027.2 | AAH19027.1 | |||
| BC095442.1 | AAH95442.1 | |||
| BG717693.1 | ||||
| BP375944.1 | ||||
| J04977.1 | AAA59475.1 | |||
| M30938.1 | AAA36154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460096 | Chr.2:216104519 - 216166605 on Build GRCh38 | Loss |
|
| nsv1011506 | Chr.2:215969945 - 216310768 on Build GRCh38 | Gain |
|
| nsv998837 | Chr.2:215982428 - 216143671 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map