Assay Details
Target Gene Details
Entrez Gene ID: | 2820 |
Gene Name: | glycerol-3-phosphate dehydrogenase 2 |
Gene Aliases: |
GDH2, GPDM, mGPDH |
Location: |
Chr.2:156435290-156586403 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPD2 | NM_001083112.2 | NP_001076581.2 | ||
| XM_011510977.2 | XP_011509279.1 | |||
| AB209399.1 | BAD92636.1 | |||
| AK290057.1 | ||||
| AK291065.1 | ||||
| AK299645.1 | ||||
| U36310.1 | AAB60403.1 | |||
| U79250.1 | AAB50200.1 |
More Information
Set Membership: |
|

Genomic Map