Assay Details
Target Gene Details
Entrez Gene ID: | 57181 |
Gene Name: | solute carrier family 39 member 10 |
Gene Aliases: |
LZT-Hs2 |
Location: |
Chr.2:195575949-195737702 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A10 | XM_011511505.2 | XP_011509807.1 |
More Information
Set Membership: |
|

Genomic Map