Assay Details
Target Gene Details
Entrez Gene ID: | 22924 |
Gene Name: | microtubule associated protein RP/EB family member 3 |
Gene Aliases: |
EB3, EBF3, EBF3-S, RP3 |
Location: |
Chr.2:26970371-27027219 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPRE3 | NM_012326.3 | NP_036458.2 | ||
| XM_006711967.3 | XP_006712030.1 | |||
| AB025186.1 | BAA82958.1 | |||
| AK312342.1 | ||||
| BC011557.1 | AAH11557.1 | |||
| DC417465.1 | ||||
| Y11174.1 | CAA72060.1 |
More Information
Set Membership: |
|

Genomic Map