Assay Details
Target Gene Details
Entrez Gene ID: | 4358 |
Gene Name: | MPV17, mitochondrial inner membrane protein |
Gene Aliases: |
MTDPS6, SYM1 |
Location: |
Chr.2:27309492-27323102 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPV17 | NM_002437.4 | NP_002428.1 | ||
| XM_005264326.3 | XP_005264383.1 | |||
| XM_006712021.3 | XP_006712084.1 | |||
| XM_017004150.1 | XP_016859639.1 | |||
| XM_017004151.1 | XP_016859640.1 | |||
| XM_017004152.1 | XP_016859641.1 | |||
| AF088036.1 | ||||
| AK093305.1 | ||||
| BC001115.2 | AAH01115.1 | |||
| BC016289.2 | AAH16289.2 | |||
| BG714998.1 | ||||
| BI602840.1 | ||||
| KU178195.1 | ||||
| KU178196.1 | ||||
| KU178197.1 | ||||
| KU178198.1 | ||||
| X76538.1 | CAA54047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
| nsv2654 | Chr.2:27309443 - 27336770 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73924414] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map