Assay Details
Target Gene Details
Entrez Gene ID: | 104326052 |
Gene Name: | negative regulator of interferon response (non-protein coding) |
Gene Aliases: |
lncRNA-CMPK2 |
Location: |
Chr.2:6828554-6840464 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRIR | NR_126359.1 | |||
| BP421873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589733 | Chr.2:6832866 - 6846168 on Build GRCh38 | Loss |
|
| esv2667897 | Chr.2:6833043 - 6845996 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74888215] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map