Assay Details
Target Gene Details
Entrez Gene ID: | 5334 |
Gene Name: | phospholipase C like 1 |
Gene Aliases: |
PLCE, PLCL, PLDL1, PPP1R127, PRIP |
Location: |
Chr.2:197804702-198149884 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCL1 | NM_006226.3 | NP_006217.3 | ||
| XM_005246643.3 | XP_005246700.1 | |||
| XM_011511351.2 | XP_011509653.1 | |||
| AK127514.1 | ||||
| AK302673.1 | ||||
| DA193701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834506 | Chr.2:197723410 - 197881428 on Build GRCh38 | Gain |
|
| nsv584157 | Chr.2:197766990 - 197848507 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map