Assay Details
Target Gene Details
Entrez Gene ID: | 55676 |
Gene Name: | solute carrier family 30 member 6 |
Gene Aliases: |
MST103, MSTP103, ZNT6 |
Location: |
Chr.2:32165841-32224806 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A6 | NM_001193513.1 | NP_001180442.1 | ||
| NM_001193514.1 | NP_001180443.1 | |||
| NM_001193515.1 | NP_001180444.1 | |||
| NM_017964.3 | NP_060434.2 | |||
| XM_006712048.3 | XP_006712111.1 | |||
| XM_011532959.2 | XP_011531261.1 | |||
| XM_011532960.2 | XP_011531262.1 | |||
| XM_011532961.2 | XP_011531263.1 | |||
| XM_011532962.2 | XP_011531264.1 | |||
| XM_011532963.2 | XP_011531265.1 | |||
| XM_017004462.1 | XP_016859951.1 | |||
| XM_017004463.1 | XP_016859952.1 | |||
| XM_017004464.1 | XP_016859953.1 | |||
| XM_017004465.1 | XP_016859954.1 | |||
| XM_017004466.1 | XP_016859955.1 | |||
| AK055663.1 | BAB70980.1 | |||
| AK096672.1 | ||||
| AK127716.1 | ||||
| AK304225.1 | ||||
| AL832755.1 | ||||
| BC066903.1 | AAH66903.1 | |||
| DB469378.1 | ||||
| EF560719.1 | ||||
| EF560726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590242 | Chr.2:32158028 - 32203815 on Build GRCh38 | Gain |
|
| nsv833670 | Chr.2:32070709 - 32256788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map