Assay Details
Target Gene Details
Entrez Gene ID: | 25801 |
Gene Name: | grancalcin |
Gene Aliases: |
GCL |
Location: |
Chr.2:162318768-162371605 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCA | NM_012198.3 | NP_036330.1 | ||
| XM_005246446.2 | XP_005246503.1 | |||
| XM_006712398.3 | XP_006712461.1 | |||
| XM_006712400.3 | XP_006712463.1 | |||
| XM_006712401.3 | XP_006712464.1 | |||
| XM_011510926.2 | XP_011509228.1 | |||
| XM_011510927.2 | XP_011509229.1 | |||
| XM_017003764.1 | XP_016859253.1 | |||
| XM_017003765.1 | XP_016859254.1 | |||
| XM_017003766.1 | XP_016859255.1 | |||
| XM_017003767.1 | XP_016859256.1 | |||
| XM_017003768.1 | XP_016859257.1 | |||
| XM_017003769.1 | XP_016859258.1 | |||
| AB209768.1 | BAD93005.1 | |||
| AK295413.1 | ||||
| AK312349.1 | ||||
| BC005214.1 | AAH05214.1 | |||
| M81637.1 | AAA58498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583520 | Chr.2:162267541 - 162611723 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map