Assay Details
Target Gene Details
Entrez Gene ID: | 51692 |
Gene Name: | cleavage and polyadenylation specific factor 3 |
Gene Aliases: |
CPSF-73, CPSF73 |
Location: |
Chr.2:9423537-9473110 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPSF3 | NM_001321833.1 | NP_001308762.1 | ||
| NM_001321834.1 | NP_001308763.1 | |||
| NM_001321835.1 | NP_001308764.1 | |||
| NM_001321836.1 | NP_001308765.1 | |||
| NM_016207.3 | NP_057291.1 | |||
| AF171877.1 | AAF00224.1 | |||
| AK223487.1 | BAD97207.1 | |||
| AK298918.1 | ||||
| BC011654.2 | AAH11654.1 | |||
| BC020211.1 | AAH20211.1 | |||
| BC030988.1 | ||||
| BG773037.1 | ||||
| DA455023.1 | ||||
| DA478774.1 | ||||
| DC420903.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain |
|
| nsv2602 | Chr.2:9408817 - 9453535 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map