Assay Details
Target Gene Details
Entrez Gene ID: | 80146 |
Gene Name: | UDP-glucuronate decarboxylase 1 |
Gene Aliases: |
SDR6E1, UGD |
Location: |
Chr.2:106093303-106194339 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524810 | Chr.2:106109472 - 106113536 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map