Assay Details
Target Gene Details
Entrez Gene ID: | 1386 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
CRE-BP1, CREB-2, CREB2, HB16, TREB7 |
Location: |
Chr.2:175072250-175168206 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012709 | Chr.2:175023400 - 175840083 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map