Assay Details
Target Gene Details
Entrez Gene ID: | 101929512 |
Gene Name: | uncharacterized LOC101929512 |
Gene Aliases: |
- |
Location: |
Chr.2:161223258-161254643 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929512 | NR_110593.1 | |||
| BX107690.1 |
Target Gene Details
Entrez Gene ID: | 10010 |
Gene Name: | TRAF family member associated NFKB activator |
Gene Aliases: |
I-TRAF, ITRAF, TRAF2 |
Location: |
Chr.2:161136955-161236176 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TANK | NM_001199135.1 | NP_001186064.1 | ||
| NM_004180.2 | NP_004171.2 | |||
| XM_005246207.3 | XP_005246264.2 | |||
| XM_017003093.1 | XP_016858582.1 | |||
| XM_017003094.1 | XP_016858583.1 | |||
| XM_017003095.1 | XP_016858584.1 | |||
| XM_017003096.1 | XP_016858585.1 | |||
| XM_017003097.1 | XP_016858586.1 | |||
| AK313096.1 | ||||
| BC067779.1 | AAH67779.1 | |||
| U59863.1 | AAC50681.1 | |||
| U63830.1 | AAC50770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7140n54 | Chr.2:161155624 - 161270523 on Build GRCh38 | Loss |
|
| nsv834437 | Chr.2:161075774 - 161268203 on Build GRCh38 | Gain |
|
| nsv437557 | Chr.2:161195062 - 161236129 on Build GRCh38 | Loss |
|
| nsv459934 | Chr.2:161203306 - 161389321 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map