Assay Details
Target Gene Details
Entrez Gene ID: | 9360 |
Gene Name: | peptidylprolyl isomerase G |
Gene Aliases: |
CARS-Cyp, CYP, SCAF10, SRCyp |
Location: |
Chr.2:169584340-169641406 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIG | NM_004792.2 | NP_004783.2 | ||
| XM_005246966.1 | XP_005247023.1 | |||
| XM_005246967.1 | XP_005247024.1 | |||
| XM_017005302.1 | XP_016860791.1 | |||
| AK307552.1 | ||||
| BC001555.1 | AAH01555.1 | |||
| BC063711.1 | AAH63711.1 | |||
| BC073875.1 | AAH73875.1 | |||
| BC109373.1 | AAI09374.1 | |||
| BC111693.1 | AAI11694.1 | |||
| U40763.1 | AAB40347.1 | |||
| X99717.1 | CAA68053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3025 | Chr.2:169609193 - 169643885 on Build GRCh38 | Deletion |
|
| nsv834454 | Chr.2:169614549 - 169774376 on Build GRCh38 | Loss |
|
| nsv1011368 | Chr.2:169423740 - 170042296 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map