Assay Details
Target Gene Details
Entrez Gene ID: | 101929452 |
Gene Name: | uncharacterized LOC101929452 |
Gene Aliases: |
- |
Location: |
Chr.2:7062727-7077880 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929452 | NR_110252.1 | |||
| DB071295.1 |
Target Gene Details
Entrez Gene ID: | 9781 |
Gene Name: | ring finger protein 144A |
Gene Aliases: |
RNF144, UBCE7IP4 |
Location: |
Chr.2:6917392-7076886 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF144A | XM_017005398.1 | XP_016860887.1 | ||
| XM_017005399.1 | XP_016860888.1 | |||
| XM_017005400.1 | XP_016860889.1 | |||
| XM_017005402.1 | XP_016860891.1 | |||
| XM_017005403.1 | XP_016860892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469394 | Chr.2:7057068 - 7090857 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map