Assay Details
Target Gene Details
Entrez Gene ID: | 65080 |
Gene Name: | mitochondrial ribosomal protein L44 |
Gene Aliases: |
COXPD16, L44MT, MRP-L44 |
Location: |
Chr.2:223943224-223967714 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL44 | XM_011511668.2 | XP_011509970.1 | ||
| XM_017004730.1 | XP_016860219.1 | |||
| XM_017004731.1 | XP_016860220.1 |
More Information
Set Membership: |
|

Genomic Map