Assay Details
Target Gene Details
Entrez Gene ID: | 90411 |
Gene Name: | multiple coagulation factor deficiency 2 |
Gene Aliases: |
F5F8D, F5F8D2, LMAN1IP, SDNSF |
Location: |
Chr.2:46901870-46941855 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCFD2 | NM_001171506.2 | NP_001164977.1 | ||
| NM_001171507.2 | NP_001164978.1 | |||
| NM_001171508.2 | NP_001164979.1 | |||
| NM_001171509.2 | NP_001164980.1 | |||
| NM_001171510.2 | NP_001164981.1 | |||
| NM_001171511.2 | NP_001164982.1 | |||
| NM_139279.5 | NP_644808.1 | |||
| AF475284.1 | AAM28465.1 | |||
| AF537214.1 | AAP23162.1 | |||
| AK292127.1 | ||||
| AK293889.1 | ||||
| AL833900.1 | CAD38756.1 | |||
| BC037845.1 | AAH37845.1 | |||
| BC040357.2 | AAH40357.1 | |||
| BI553070.1 | ||||
| CF994192.1 | ||||
| CR749562.1 | CAH18359.1 | |||
| DA194356.1 | ||||
| DA226366.1 | ||||
| DA324623.1 | ||||
| DA330030.1 | ||||
| DA404031.1 | ||||
| DA412975.1 | ||||
| DB096245.1 | ||||
| DC321899.1 | ||||
| DC332658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590618 | Chr.2:46911622 - 46913607 on Build GRCh38 | Loss |
|
| esv3590617 | Chr.2:46911358 - 46916995 on Build GRCh38 | Loss |
|
| esv2677617 | Chr.2:46911650 - 46913648 on Build GRCh38 | Deletion |
|
| nsv457352 | Chr.2:46777649 - 47098834 on Build GRCh38 | Loss |
|
| nsv1004828 | Chr.2:46829710 - 47587343 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map