Assay Details
Target Gene Details
Entrez Gene ID: | 3336 |
Gene Name: | heat shock protein family E (Hsp10) member 1 |
Gene Aliases: |
CPN10, EPF, GROES, HSP10 |
Location: |
Chr.2:197499997-197503463 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPE1 | NM_002157.2 | NP_002148.1 | ||
| AK312104.1 | ||||
| BC023518.2 | AAH23518.1 | |||
| BG188453.1 | ||||
| CN264788.1 | ||||
| CR407688.1 | CAG28616.1 | |||
| U07550.1 | AAA50953.1 | |||
| X75821.1 | CAA53455.1 |
Target Gene Details
Entrez Gene ID: | 100529241 |
Gene Name: | HSPE1-MOB4 readthrough |
Gene Aliases: |
HSPE1-PHOCN |
Location: |
Chr.2:197499997-197553699 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPE1-MOB4 | NM_001202485.1 | NP_001189414.1 | ||
| BJ992310.1 | ||||
| CR990871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014881 | Chr.2:197432432 - 197668882 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map