Assay Details
Target Gene Details
Entrez Gene ID: | 6744 |
Gene Name: | sperm specific antigen 2 |
Gene Aliases: |
CS-1, CS1, KRAP, SPAG13 |
Location: |
Chr.2:181891716-181930738 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSFA2 | NM_001130445.2 | NP_001123917.1 | ||
| NM_001287503.1 | NP_001274432.1 | |||
| NM_001287504.1 | NP_001274433.1 | |||
| NM_006751.6 | NP_006742.2 | |||
| NR_109843.1 | ||||
| XM_005246812.1 | XP_005246869.1 | |||
| XM_005246813.1 | XP_005246870.1 | |||
| XM_011511701.2 | XP_011510003.1 | |||
| XM_017004782.1 | XP_016860271.1 | |||
| AB067514.2 | ||||
| AB116937.1 | BAC81767.1 | |||
| AK291745.1 | ||||
| AK304238.1 | ||||
| BC028706.2 | AAH28706.1 | |||
| BC052581.1 | AAH52581.1 | |||
| BC064499.1 | AAH64499.1 | |||
| BX647257.1 | ||||
| BX647967.1 | ||||
| BX648888.1 | ||||
| CR749488.1 | CAH18314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834479 | Chr.2:181873756 - 182040717 on Build GRCh38 | Gain |
|
| esv3593551 | Chr.2:181846661 - 181930879 on Build GRCh38 | Gain |
|
| esv3593550 | Chr.2:181846220 - 181983097 on Build GRCh38 | Gain |
|
| nsv834478 | Chr.2:181790084 - 181978474 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map