Assay Details
Target Gene Details
Entrez Gene ID: | 150737 |
Gene Name: | tetratricopeptide repeat domain 30B |
Gene Aliases: |
IFT70, IFT70B, fleer |
Location: |
Chr.2:177550152-177552796 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC30B | NM_152517.2 | NP_689730.2 | ||
| AK055552.1 | BAB70953.1 | |||
| BC033795.1 | AAH33795.1 | |||
| DA260391.1 | ||||
| DA557876.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv961534 | Chr.2:177550976 - 177552881 on Build GRCh38 | Duplication |
|
| esv2762964 | Chr.2:177551666 - 177617090 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map