Assay Details
Target Gene Details
Entrez Gene ID: | 4719 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S1 |
Gene Aliases: |
CI-75Kd, CI-75k, PRO1304 |
Location: |
Chr.2:206121971-206159519 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFS1 | XM_017004188.1 | XP_016859677.1 | ||
| BC030833.1 | AAH30833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893538 | Chr.2:206115621 - 206147759 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map