Assay Details
Target Gene Details
Entrez Gene ID: | 55233 |
Gene Name: | MOB kinase activator 1A |
Gene Aliases: |
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B |
Location: |
Chr.2:74152598-74178967 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB1A | NM_001317110.1 | 6 | 2847 | NP_001304039.1 |
| NM_001317111.1 | 6 | 2743 | NP_001304040.1 | |
| NM_001317112.1 | 5 | 2686 | NP_001304041.1 | |
| NM_018221.4 | 6 | 2850 | NP_060691.2 | |
| XM_011532948.2 | 5 | 2546 | XP_011531250.1 | |
| AL122062.1 | 1 | 565 |
More Information
Set Membership: |
|

Genomic Map