Assay Details
Target Gene Details
Entrez Gene ID: | 104326052 |
Gene Name: | negative regulator of interferon response (non-protein coding) |
Gene Aliases: |
lncRNA-CMPK2 |
Location: |
Chr.2:6828554-6840464 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRIR | NR_126359.1 | |||
| BP421873.1 | ||||
| BQ437827.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77588481] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map