Assay Details
Target Gene Details
Entrez Gene ID: | 94101 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189766149-189784371 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORMDL1 | XM_011512199.2 | 4 | 507 | XP_011510501.1 |
| XM_017005348.1 | 4 | 496 | XP_016860837.1 |
More Information
Set Membership: |
|

Genomic Map