Assay Details
Target Gene Details
Entrez Gene ID: | 238 |
Gene Name: | anaplastic lymphoma receptor tyrosine kinase |
Gene Aliases: |
CD246, NBLST3 |
Location: |
Chr.2:29192774-29921611 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALK | NM_004304.4 | NP_004295.2 | ||
| AB209477.4 | BAD92714.1 | |||
| AK296218.1 | ||||
| EU788003.1 | ||||
| EU788004.1 | ||||
| EU788005.1 | ||||
| EU788006.1 | ||||
| EU788007.1 | ||||
| EU788008.1 | ||||
| EU788009.1 | ||||
| EU788010.1 | ||||
| FJ838786.1 | ||||
| FJ838787.1 | ||||
| GU128152.1 | ||||
| GU128153.1 | ||||
| GU128154.1 | ||||
| GU128155.1 | ||||
| GU128156.1 | ||||
| GU128157.1 | ||||
| U62540.1 | AAB71619.1 | |||
| U66559.1 | AAC51104.1 |
Target Gene Details
Entrez Gene ID: | 105374389 |
Gene Name: | uncharacterized LOC105374389 |
Gene Aliases: |
- |
Location: |
Chr.2:29474734-29475377 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105374389 | NR_136317.1 | 2 | 259 | |
| AA448482.1 | 1 | 148 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34904 | Chr.2:28979483 - 30284483 on Build GRCh38 | Gain |
|
| nsv1006359 | Chr.2:29459991 - 29501317 on Build GRCh38 | Loss |
|
| nsv1008490 | Chr.2:28884768 - 29482018 on Build GRCh38 | Gain |
|
| esv2759035 | Chr.2:28953630 - 30316563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map