Assay Details
Target Gene Details
Entrez Gene ID: | 10018 |
Gene Name: | BCL2 like 11 |
Gene Aliases: |
BAM, BIM, BOD |
Location: |
Chr.2:111120914-111168445 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L11 | NM_001204106.1 | 1 | 182 | NP_001191035.1 |
| NM_001204107.1 | 1 | 182 | NP_001191036.1 | |
| NM_001204108.1 | 1 | 182 | NP_001191037.1 | |
| NM_001204109.1 | 1 | 182 | NP_001191038.1 | |
| NM_001204110.1 | 1 | 182 | NP_001191039.1 | |
| NM_001204111.1 | 1 | 182 | NP_001191040.1 | |
| NM_001204112.1 | 1 | 182 | NP_001191041.1 | |
| NM_001204113.1 | 1 | 182 | NP_001191042.1 | |
| NM_006538.4 | 1 | 182 | NP_006529.1 | |
| NM_138621.4 | 1 | 182 | NP_619527.1 | |
| NM_138622.3 | 1 | 182 | NP_619528.1 | |
| NM_138623.3 | 1 | 182 | NP_619529.1 | |
| NM_138624.3 | 1 | 182 | NP_619530.1 | |
| NM_138625.3 | 1 | 182 | NP_619531.1 | |
| NM_138626.3 | 1 | 182 | NP_619532.1 | |
| NM_138627.3 | 1 | 182 | NP_619533.1 | |
| NM_207002.3 | 1 | 182 | NP_996885.1 | |
| NM_207003.2 | 1 | 182 | NP_996886.1 | |
| XM_005263556.3 | 1 | 182 | XP_005263613.1 | |
| XM_005263561.3 | 1 | 182 | XP_005263618.1 | |
| XM_017003102.1 | 1 | 182 | XP_016858591.1 | |
| AK290377.1 | 1 | 182 | ||
| AK291269.1 | 1 | 182 | ||
| BC033694.1 | 1 | 167 | AAH33694.1 | |
| CN411912.1 | 1 | 152 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829618 | Chr.2:111118050 - 111123970 on Build GRCh38 | Gain |
|
| nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss |
|
| nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss |
|
| nsv834331 | Chr.2:111054562 - 111247101 on Build GRCh38 | Loss |
|
| dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map