Assay Details
Target Gene Details
Entrez Gene ID: | 4664 |
Gene Name: | NGFI-A binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:190648860-190692766 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584252 | Chr.2:190654887 - 190674560 on Build GRCh38 | Loss |
|
| nsv3082 | Chr.2:190627513 - 190688068 on Build GRCh38 | Insertion |
|
| nsv834492 | Chr.2:190574138 - 190781895 on Build GRCh38 | Gain |
|
| dgv1200e212 | Chr.2:190638342 - 190674560 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map