Assay Details
Target Gene Details
Entrez Gene ID: | 56902 |
Gene Name: | partner of NOB1 homolog |
Gene Aliases: |
KHRBP1, RRP20 |
Location: |
Chr.2:68157873-68175959 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNO1 | NM_020143.2 | NP_064528.1 | ||
| AF164799.1 | AAF80763.1 | |||
| AF349314.1 | AAK29434.1 | |||
| AK223118.1 | BAD96838.1 | |||
| AK225641.1 | ||||
| AK291715.1 | ||||
| AK313204.1 | ||||
| BC008304.1 | AAH08304.1 | |||
| BC018152.1 | AAH18152.1 | |||
| L48692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834251 | Chr.2:68015596 - 68200603 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map