Assay Details
Target Gene Details
Entrez Gene ID: | 4862 |
Gene Name: | neuronal PAS domain protein 2 |
Gene Aliases: |
MOP4, PASD4, bHLHe9 |
Location: |
Chr.2:100820151-100996829 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPAS2 | NM_002518.3 | NP_002509.2 | ||
| XM_005263953.2 | XP_005264010.1 | |||
| XM_005263959.2 | XP_005264016.1 | |||
| XM_005263960.2 | XP_005264017.1 | |||
| XM_011511242.2 | XP_011509544.1 | |||
| XM_011511243.2 | XP_011509545.1 | |||
| XM_017004214.1 | XP_016859703.1 | |||
| XM_017004215.1 | XP_016859704.1 | |||
| XM_017004216.1 | XP_016859705.1 | |||
| XM_017004217.1 | XP_016859706.1 | |||
| AK294733.1 | ||||
| BC051351.1 | AAH51351.2 | |||
| BC072383.1 | AAH72383.1 | |||
| U77970.1 | AAB47250.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582532 | Chr.2:100412540 - 100915481 on Build GRCh38 | Gain |
|
| nsv829584 | Chr.2:100814389 - 100823820 on Build GRCh38 | Gain |
|
| nsv834315 | Chr.2:100768034 - 100932968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map