Assay Details
Target Gene Details
Entrez Gene ID: | 64965 |
Gene Name: | mitochondrial ribosomal protein S9 |
Gene Aliases: |
MRP-S9, RPMS9, S9mt |
Location: |
Chr.2:105038025-105099960 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS9 | NM_182640.2 | NP_872578.1 | ||
| AK001451.1 | ||||
| AK130078.1 | ||||
| AV729381.1 | ||||
| BC057240.1 | AAH57240.1 | |||
| DT217261.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77392827] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map