Assay Details
Target Gene Details
Entrez Gene ID: | 51072 |
Gene Name: | mediator of cell motility 1 |
Gene Aliases: |
C2orf4, CGI-27, MEMO, NS5ATP7 |
Location: |
Chr.2:31867188-32011052 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEMO1 | NM_001137602.2 | NP_001131074.1 | ||
| NM_001301833.1 | NP_001288762.1 | |||
| NM_001301852.1 | NP_001288781.1 | |||
| NM_015955.3 | NP_057039.1 | |||
| NR_126032.1 | ||||
| NR_126034.1 | ||||
| XM_011532892.2 | XP_011531194.1 | |||
| XM_011532896.2 | XP_011531198.1 | |||
| XM_017004255.1 | XP_016859744.1 | |||
| AB041018.1 | BAD74066.1 | |||
| AB041019.1 | BAD74067.1 | |||
| AF132961.1 | AAD27736.1 | |||
| AF363446.1 | AAL34462.1 | |||
| AF529368.1 | AAQ09602.1 | |||
| AK057760.1 | ||||
| AK290753.1 | ||||
| AK297128.1 | ||||
| BC018733.1 | AAH18733.1 | |||
| BC070046.1 | AAH70046.1 | |||
| BC094681.1 | AAH94681.1 | |||
| BG033287.1 | ||||
| CD250945.1 |
More Information
Set Membership: |
|

Genomic Map