Assay Details
Target Gene Details
Entrez Gene ID: | 8575 |
Gene Name: | protein activator of interferon induced protein kinase EIF2AK2 |
Gene Aliases: |
DYT16, HSD14, PACT, RAX |
Location: |
Chr.2:178431414-178451240 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKRA | NM_001139517.1 | NP_001132989.1 | ||
| NM_001139518.1 | NP_001132990.1 | |||
| NM_001316362.1 | NP_001303291.1 | |||
| NM_003690.4 | NP_003681.1 | |||
| XM_011512063.1 | XP_011510365.1 | |||
| XM_011512066.1 | XP_011510368.1 | |||
| XM_017005159.1 | XP_016860648.1 | |||
| AF072860.1 | AAC25672.1 | |||
| AF083033.1 | AAD33099.1 | |||
| AK223107.1 | BAD96827.1 | |||
| AK290601.1 | ||||
| AK296019.1 | ||||
| AL136615.1 | CAB66550.1 | |||
| AL833867.1 | CAD38725.1 | |||
| AY251164.1 | AAP20061.1 | |||
| BC009470.1 | AAH09470.1 | |||
| BG827125.1 | ||||
| BT007243.1 | AAP35907.1 | |||
| CR533525.1 | CAG38556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv9296 | Chr.2:178443315 - 178450320 on Build GRCh38 | Loss |
|
| esv2721193 | Chr.2:178443386 - 178450224 on Build GRCh38 | Deletion |
|
| nsv834467 | Chr.2:178393413 - 178566238 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map