Assay Details
Target Gene Details
Entrez Gene ID: | 26146 |
Gene Name: | TRAF3 interacting protein 1 |
Gene Aliases: |
IFT54, MIP-T3, MIPT3, SLSN9 |
Location: |
Chr.2:238320489-238400900 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3IP1 | NM_001139490.1 | NP_001132962.1 | ||
| NM_015650.3 | NP_056465.2 | |||
| XM_006712414.2 | XP_006712477.1 | |||
| XM_011510944.2 | XP_011509246.1 | |||
| XM_011510945.2 | XP_011509247.1 | |||
| XM_011510946.2 | XP_011509248.1 | |||
| XM_011510947.2 | XP_011509249.1 | |||
| XM_011510948.2 | XP_011509250.1 | |||
| XM_017003789.1 | XP_016859278.1 | |||
| AF230877.1 | AAF76984.1 | |||
| AF242456.1 | AAL90444.1 | |||
| BC059174.1 | AAH59174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3233 | Chr.2:238303159 - 238335494 on Build GRCh38 | Insertion |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75498923] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map