Assay Details
Target Gene Details
Entrez Gene ID: | 129607 |
Gene Name: | cytidine/uridine monophosphate kinase 2 |
Gene Aliases: |
NDK, TMPK2, TYKi, UMP-CMPK2 |
Location: |
Chr.2:6840417-6866635 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMPK2 | NM_001256477.1 | 4 | 1982 | NP_001243406.1 |
| NM_001256478.1 | NP_001243407.1 | |||
| NM_207315.3 | NP_997198.2 | |||
| NR_046236.1 | ||||
| AK127983.1 | BAC87217.1 | |||
| BC016969.1 | AAH16969.1 | |||
| BC089425.1 | AAH89425.1 | |||
| BC132821.1 | ||||
| BC141802.1 | 4 | 1875 | ||
| BC144202.1 | ||||
| BX463706.2 | ||||
| DA501203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833326 | Chr.2:6844896 - 7002433 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map