Assay Details
Target Gene Details
Entrez Gene ID: | 65080 |
Gene Name: | mitochondrial ribosomal protein L44 |
Gene Aliases: |
COXPD16, L44MT, MRP-L44 |
Location: |
Chr.2:223943224-223967714 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL44 | NM_022915.3 | NP_075066.1 | ||
| XM_011511668.2 | XP_011509970.1 | |||
| XM_017004730.1 | XP_016860219.1 | |||
| XM_017004731.1 | XP_016860220.1 | |||
| AK022763.1 | BAB14234.1 | |||
| AK024052.1 | BAB14802.1 | |||
| AK095007.1 | 1 | 1937 | ||
| BC012058.1 | AAH12058.1 | |||
| CR457293.1 | CAG33574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv511173 | Chr.2:223964259 - 223970652 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map