Assay Details
Target Gene Details
Entrez Gene ID: | 54842 |
Gene Name: | major facilitator superfamily domain containing 6 |
Gene Aliases: |
MMR2, hMMR2 |
Location: |
Chr.2:190408355-190502315 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD6 | NM_017694.3 | NP_060164.3 | ||
| XM_005246654.2 | XP_005246711.1 | |||
| XM_005246656.3 | XP_005246713.1 | |||
| XM_011511368.1 | XP_011509670.1 | |||
| XM_017004392.1 | XP_016859881.1 | |||
| AK127176.1 | BAC86869.1 |

Genomic Map