Assay Details
Target Gene Details
Entrez Gene ID: | 90134 |
Gene Name: | potassium voltage-gated channel subfamily H member 7 |
Gene Aliases: |
ERG3, HERG3, Kv11.3 |
Location: |
Chr.2:162371407-162838747 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH7 | NM_033272.3 | NP_150375.2 | ||
| XM_011512109.2 | XP_011510411.1 | |||
| XM_017005218.1 | XP_016860707.1 | |||
| XM_017005219.1 | XP_016860708.1 | |||
| XM_017005220.1 | XP_016860709.1 | |||
| AF032897.1 | AAD01946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583520 | Chr.2:162267541 - 162611723 on Build GRCh38 | Loss |
|
| nsv1160878 | Chr.2:162355987 - 162425533 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map