Assay Details
Target Gene Details
Entrez Gene ID: | 83879 |
Gene Name: | cell division cycle associated 7 |
Gene Aliases: |
ICF3, JPO1 |
Location: |
Chr.2:173354017-173368992 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA7 | XM_011511957.2 | XP_011510259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834462 | Chr.2:173327577 - 173506798 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map