Assay Details
Target Gene Details
Entrez Gene ID: | 64965 |
Gene Name: | mitochondrial ribosomal protein S9 |
Gene Aliases: |
MRP-S9, RPMS9, S9mt |
Location: |
Chr.2:105038025-105099960 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS9 | NM_182640.2 | NP_872578.1 | ||
| AK001451.1 | ||||
| AK130078.1 | ||||
| AV729381.1 | ||||
| BC057240.1 | AAH57240.1 | |||
| DT217261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6983n54 | Chr.2:105046454 - 105049076 on Build GRCh38 | Loss |
|
| esv3591824 | Chr.2:105045653 - 105049184 on Build GRCh38 | Loss |
|
| esv3559884 | Chr.2:105045543 - 105049144 on Build GRCh38 | Deletion |
|
| esv27425 | Chr.2:105045585 - 105049099 on Build GRCh38 | Loss |
|
| esv2663750 | Chr.2:105045645 - 105049130 on Build GRCh38 | Deletion |
|
| nsv582576 | Chr.2:105047202 - 105049350 on Build GRCh38 | Loss |
|
| dgv85e180 | Chr.2:105045528 - 105049124 on Build GRCh38 | Loss |
|
| dgv86e180 | Chr.2:105045538 - 105050067 on Build GRCh38 | Deletion |
|
| esv2842792 | Chr.2:105045649 - 105049134 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map